Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78519

Sigma-Aldrich

Anti-Rabbit IgG - Atto 655 antibody produced in goat

~1 mg/mL IgG

Synonym(s):

Atto 655 [goat-Anti-rabbit IgG]

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

Atto 655 conjugate

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

glycerol

species reactivity

rabbit

concentration

~1 mg/mL IgG

fluorescence

λex 655 nm; λem 679 nm in PBS

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgGs are glycoprotein antibodies that modulate several immune responses. Rabbit IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rabbit IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins. Goat anti-rabbit IgGs are known to associate with rabbit IgGs.

Immunogen

rabbit IgG

Application

Atto 655-goat anti-rabbit IgG may be used for fluorescence-based immunoassays.

Analysis Note

Unconjugated dye ≤5% of total fluorescence; dye-to-protein ratio ≥2

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service