Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78438

Sigma-Aldrich

Dy-615 NHS ester

suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H41N3O8S
Molecular Weight:
675.79
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

form

powder

fluorescence

λex 610 nm; λem 631 nm in 0.1 M phosphate pH 7.0

suitability

corresponds for coupling to amines
suitable for fluorescence

storage temp.

2-8°C

SMILES string

CC(C)(C)c1cc(\C=C\C=C2\N(CCCCCC(=O)ON3C(=O)CCC3=O)c4ccc(cc4C2(C)C)S([O-])(=O)=O)c5ccc(N)cc5[o+]1

InChI

1S/C36H41N3O8S/c1-35(2,3)31-20-23(26-15-13-24(37)21-29(26)46-31)10-9-11-30-36(4,5)27-22-25(48(43,44)45)14-16-28(27)38(30)19-8-6-7-12-34(42)47-39-32(40)17-18-33(39)41/h9-11,13-16,20-22H,6-8,12,17-19,37H2,1-5H3/b10-9+,30-11+

InChI key

NIOGLMJYJBEELX-GINSVCLMSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuliya Voskobiynyk et al.
PLoS genetics, 17(1), e1009195-e1009195 (2021-01-08)
Dravet syndrome (DS) is a developmental and epileptic encephalopathy that results from mutations in the Nav1.1 sodium channel encoded by SCN1A. Most known DS-causing mutations are in coding regions of SCN1A, but we recently identified several disease-associated SCN1A mutations in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service