Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78319

Sigma-Aldrich

trans-Glutaconic acid sodium salt hydrate

≥95.0% (HPLC)

Synonym(s):

(2E)-Pentenedioic acid sodium salt hydrate, Sodium trans-glutaconate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O4 · xNa+ · yH2O
Molecular Weight:
130.10 (anhydrous free acid basis)
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.25

assay

≥95.0% (HPLC)

form

powder

SMILES string

OC(C/C=C/C(O)=O)=O.C

InChI

1S/C5H6O4.CH4/c6-4(7)2-1-3-5(8)9;/h1-2H,3H2,(H,6,7)(H,8,9);1H4/b2-1+;

InChI key

NRJOWSDIBMLXCK-TYYBGVCCSA-N

Application

Internal reference for urine samples

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atsuki Ikeda et al.
Biomedical chromatography : BMC, 26(5), 548-558 (2011-07-21)
Conventional tumor markers are unsuitable for detecting carcinoma at an early stage and lack clinical efficacy and utility. In this study, we attempted to investigate the differences in serum metabolite profiles of gastrointestinal cancers and healthy volunteers using a metabolomic
G F Hoffmann et al.
Journal of inherited metabolic disease, 22(4), 381-391 (1999-07-17)
The biochemical hallmark of glutaric aciduria type I (GA I) due to glutaryl-CoA dehydrogenase deficiency is the accumulation of glutaric acid, and to a lesser degree of 3-hydroxyglutaric and glutaconic acids. Abnormal metabolites vary from gross organic aciduria to only
Metabolomics of urinary organic acids in respiratory chain deficiencies in children.
Reinecke, C.J., et al.
Metabolomics, 8, 264-283 (2012)
Júlia Dénes et al.
Analytical chemistry, 84(22), 10113-10120 (2012-10-26)
A novel, single stage high resolution mass spectrometry-based method is presented for the population level screening of inborn errors of metabolism. The approach proposed here extends traditional electrospray tandem mass spectrometry screening by introducing nanospray ionization and high resolution mass
Jonathan C Schisler et al.
Journal of the American Heart Association, 4(2), doi:10-doi:10 (2015-02-26)
Long chain acyl-CoA synthetases (ACSL) catalyze long-chain fatty acids (FA) conversion to acyl-CoAs. Temporal ACSL1 inactivation in mouse hearts (Acsl1(H-/-)) impaired FA oxidation and dramatically increased glucose uptake, glucose oxidation, and mTOR activation, resulting in cardiac hypertrophy. We used unbiased

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service