Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78302

Sigma-Aldrich

InP/ZnS nanoparticles

core-shell type, red emitting, ~1 mg/mL

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

form

liquid

concentration

~1 mg/mL

fluorescence

λem 630 nm±20 nm

Looking for similar products? Visit Product Comparison Guide

Analysis Note

full width at half maximum ≤100 nm

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

39.2 °F - closed cup - Solvent

flash_point_c

4.0 °C - closed cup - Solvent

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service