Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78105

Sigma-Aldrich

N-(L-Phenylalanyl)-2-aminoacridone

suitable for fluorescence, ≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19N3O2
CAS Number:
Molecular Weight:
357.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

assay

≥97.0% (HPLC)

form

powder

extent of labeling

0.7-1.1 mmol/g loading

particle size

≥45 μm (95%)

fluorescence

λex 398 nm; λem 440 nm in 0.1 M Tris pH 8.0

suitability

suitable for fluorescence

SMILES string

N[C@@H](Cc1ccccc1)C(=O)Nc2ccc3Nc4ccccc4C(=O)c3c2

InChI

1S/C22H19N3O2/c23-18(12-14-6-2-1-3-7-14)22(27)24-15-10-11-20-17(13-15)21(26)16-8-4-5-9-19(16)25-20/h1-11,13,18H,12,23H2,(H,24,27)(H,25,26)/t18-/m0/s1

InChI key

WINNAKHIQUIPDH-SFHVURJKSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

In addition to the emission maximum at 440 nm, there is a lower maximum at 463 nm.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service