cleaning products cosmetics flavors and fragrances food and beverages personal care
storage temp.
2-8°C
General description
filtered through a 0.45 μm membrane filter
Application
suitable as IEF-Marker for HPCE and for IEF-gels
Reconstitution
For use in HPCE the marker should be diluted 1:100 in a suitable ampholyte solution (e.g. 2% ampholyte 3-10), UV-detection at 280 nm (20 °C). For use in IEF-gels the solution can be loaded directly onto the gel
Information on Isoelectric Focusing including what it is and how it is used. In order to ensure the high performance of analysis, isoelectric point (pI) standards are needed.
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.