Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76712

Sigma-Aldrich

L-Rhamnono-1,4-lactone

≥98% (TLC)

Synonym(s):

6-Deoxy-L-mannonic acid γ-lactone, L-Rhamnonic 1,4-lactone, L-Rhamnonic acid γ-lactone, L-Rhamno-1,4-lactone, NSC 51238

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O5
CAS Number:
Molecular Weight:
162.14
Beilstein/REAXYS Number:
82278
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

assay

≥98% (TLC)

form

powder

optical activity

[α]/D -39±3°, c = 2 in H2O

storage temp.

2-8°C

SMILES string

O[C@H]([C@H]([C@@H](O)C)O1)[C@@H](O)C1=O

InChI

1S/C6H10O5/c1-2(7)5-3(8)4(9)6(10)11-5/h2-5,7-9H,1H3/t2-,3-,4+,5-/m0/s1

InChI key

VASLEPDZAKCNJX-KLVWXMOXSA-N

Biochem/physiol Actions

Metabolite of L-rhamnose catabolic pathway.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidative pathway for L-rhamnose degradation in Pullularia pullulans.
Rigo, L.U., et al.
Canadian Journal of Microbiology, 31, 817-822 (1985)
Suk Min Kim et al.
Extremophiles : life under extreme conditions, 16(3), 447-454 (2012-04-07)
Thermoplasma acidophilum utilizes L-rhamnose as a sole carbon source. To determine the metabolic pathway of L-rhamnose in Archaea, we identified and characterized L-rhamnose dehydrogenase (RhaD) in T. acidophilum. Ta0747P gene, which encodes the putative T. acidophilum RhaD (Ta_RhaD) enzyme belonging
Oxetane cis- and trans-?β-amino acid scaffolds from L-rhamnose by efficient SN2 reactions in oxetane rings; pseudoenantiomeric analogues of the antibiotic oxetin.
Johnson, S.W., et al.
Tetrahedron Asymmetry, 15, 2681-2686 (2004)
L-Rhamnose metabolism in Pichia stipitis and Debaryomyces polymorphus.
Twerdochlib, A.L., et al.
Canadian Journal of Microbiology, 40, 896-902 (1994)
Merlin Eric Hobbs et al.
Biochemistry, 52(1), 239-253 (2012-12-12)
A member of the amidohydrolase superfamily, BmulJ_04915 from Burkholderia multivorans, of unknown function was determined to hydrolyze a series of sugar lactones: L-fucono-1,4-lactone, D-arabino-1,4-lactone, L-xylono-1,4-lactone, D-lyxono-1,4-lactone, and L-galactono-1,4-lactone. The highest activity was shown for L-fucono-1,4-lactone with a k(cat) value of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service