Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76488

Sigma-Aldrich

(S)-(+)-4-(3-Amino-pyrrolidino)-7-nitrobenzofurazan

BioReagent, suitable for fluorescence, ≥98.0% (HPLC)

Synonym(s):

(S)-(+)-1-(7-Nitro-4-benzofurazanyl)-3-pyrrolidinamine, (S)-(+)-NBD-Apy

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N5O3
CAS Number:
Molecular Weight:
249.23
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

product line

BioReagent

assay

≥98.0% (HPLC)

form

powder

optical activity

[α]20/D +13.6±2°

mp

178-184 °C

fluorescence

λex ≥490 nm; λem ≥535 nm in acetonitrile

suitability

suitable for fluorescence

storage temp.

2-8°C

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service