Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75618

Sigma-Aldrich

Vitamin K2 2,3-epoxide

≥98.0% (HPLC)

Synonym(s):

1a,7a-Dihydro-1a-methyl-7a-[(2E,6E,10E)-3,7,11,15-tetramethyl-2,6,10,14-hexadecatetraen-1-yl]naphth[2,3-b]oxirene-2,7-dione, 2,3-Epoxymenaquinone, Menaquinone-4-epoxide, Menatetrenone epoxide, Menatetrenone-2,3-epoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H40O3
CAS Number:
Molecular Weight:
460.65
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.79

assay

≥98.0% (HPLC)

form

liquid

color

colorless to green-yellow

storage temp.

−20°C

SMILES string

CC1(C2(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CCC=C(C)C)O1)C(C3=CC=CC=C3C2=O)=O

InChI

1S/C31H40O3/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-31-29(33)27-19-8-7-18-26(27)28(32)30(31,6)34-31/h7-8,12,14,16,18-20H,9-11,13,15,17,21H2,1-6H3/b23-14+,24-16+,25-20+

InChI key

TUZHANAISKEZFG-GHDNBGIDSA-N

Biochem/physiol Actions

Vitamin K2 2,3-epoxide is a vitamin K metabolite occurring in the ubiquinone, secondary metabolite and other terpenoid-quinone biosynthetic pathways. Vitamin K is needed for the post-translational modification of certain proteins, mostly required for blood coagulation. Within the cell, vitamin K undergoes electron reduction to a reduced form of vitamin K by the enzyme vitamin K epoxide reductase. Another enzyme then oxidizes vitamin K hydroquinone to allow carboxylation of glutamate to gamma-carboxyglutamate (Gla); this enzyme is called the gamma-glutamyl carboxylase or the vitamin K-dependent carboxylase. The carboxylation reaction will only proceed if the carboxylase enzyme is able to oxidize vitamin K hydroquinone to vitamin K epoxide at the same time; the carboxylation and epoxidation reactions are said to be coupled reactions. Vitamin K epoxide is then re-converted to vitamin K by the vitamin K epoxide reductase. These two enzymes comprise the so-called vitamin K cycle. One of the reasons why vitamin K is rarely deficient in a human diet is because vitamin K is continually recycled in our cells. Vitamin K 2,3-epoxide is the substrate for vitamin K 2,3-epoxide reductase (VKOR) complex. Significantly increased level of serum vitamin K epoxide has been found in patients with familial multiple coagulation factor deficiency.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Miyake et al.
Osteoporosis international : a journal established as result of cooperation between the European Foundation for Osteoporosis and the National Osteoporosis Foundation of the USA, 12(8), 680-687 (2001-10-03)
It has been reported that vitamin K2 (menaquinone-4) promoted 1,25-dihydroxyvitamin D3 (1,25(OH)2D3)-induced mineralization and enhanced gamma-carboxyglutamic acid (Gla)-containing osteocalcin accumulation in cultured human osteoblasts. In the present study, we investigated whether menaquinone-4 (MK-4) was metabolized in human osteoblasts to act
Satoki Karasawa et al.
Molecular pharmacology, 83(3), 613-620 (2012-12-12)
Vitamin K2 (VK2, menaquinone) is known to have anticancer activity in vitro and in vivo. Although its effect is thought to be mediated, at least in part, by the induction of apoptosis, the underlying molecular mechanism remains elusive. Here, we
Philipp Westhofen et al.
The Journal of biological chemistry, 286(17), 15085-15094 (2011-03-04)
Human vitamin K 2,3-epoxide reductase complex subunit 1-like 1 (VKORC1L1), expressed in HEK 293T cells and localized exclusively to membranes of the endoplasmic reticulum, was found to support both vitamin K 2,3-epoxide reductase (VKOR) and vitamin K reductase enzymatic activities.
Andreas Fregin et al.
Blood, 100(9), 3229-3232 (2002-10-18)
Familial multiple coagulation factor deficiency (FMFD) of factors II, VII, IX, X, protein C, and protein S is a very rare bleeding disorder with autosomal recessive inheritance. The phenotypic presentation is variable with respect to the residual activities of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service