Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75577

Sigma-Aldrich

Lipase from Burkholderia sp.

lyophilized powder, slightly beige, ~12 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

biological source

Burkholderia spp.

form

lyophilized powder

specific activity

~12 U/mg

color

slightly beige

storage temp.

−20°C

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol oleic acid per minute at pH 8.0 and 40°C (triolein, Cat. No. 62314 as substrate)

Analysis Note

partially soluble in water or buffer

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Hall et al.
The Journal of small animal practice, 54(5), 275-279 (2013-02-13)
Mushroom toxicosis is rarely diagnosed in dogs and is poorly reported in the veterinary literature. This report suggests that mushroom toxicosis is a potentially under-diagnosed condition in first opinion practice in the UK. Nine dogs with clinical signs consistent with
Minshan Hu et al.
Advances in clinical and experimental medicine : official organ Wroclaw Medical University, 21(6), 751-757 (2013-03-05)
Diet induces changes in plasma lipid profiles, and the plasma lipid profiles vary among different genetic backgrounds. The aim of this study was to investigate how a high-carbohydrate (high-CHO) diet interacts with hepatic lipase G-250A promoter polymorphism to affect the
Stephen G Young et al.
Genes & development, 27(5), 459-484 (2013-03-12)
All organisms use fatty acids (FAs) for energy substrates and as precursors for membrane and signaling lipids. The most efficient way to transport and store FAs is in the form of triglycerides (TGs); however, TGs are not capable of traversing
Mrinalini Muralidharan et al.
Plant molecular biology, 81(6), 565-576 (2013-02-23)
Acetylcholinesterase is an enzyme that is intimately associated with regulation of synaptic transmission in the cholinergic nervous system and in neuromuscular junctions of animals. However the presence of cholinesterase activity has been described also in non-metazoan organisms such as slime
Mehran Howaizi et al.
Acta gastro-enterologica Belgica, 75(4), 446-447 (2013-02-14)
Acute pancreatitis has a large number of causes. Major causes are alcohol and gallstones. Toxic causes, mainly represented by medication-induced pancreatitis account for less than 2% of the cases. Cannabis is an anecdotally reported cause of acute pancreatitis. Six cases

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service