Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75469

Sigma-Aldrich

L-Ornithine monohydrochloride

≥99.5% (AT), BioXtra

Synonym(s):

(S)-2,5-Diaminopentanoic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2O2 · HCl
CAS Number:
Molecular Weight:
168.62
Beilstein/REAXYS Number:
3625847
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Ornithine monohydrochloride, BioXtra, ≥99.5% (AT)

product line

BioXtra

assay

≥99.5% (AT)

form

powder

optical activity

[α]20/D +23.0±0.5°, c = 5% in 5 M HCl

impurities

insoluble matter, passes filter test
≤0.3% foreign amino acids

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying, 20 °C (HV)

color

white

pH

5.0-6.0 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤200 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.06
λ: 280 nm Amax: 0.06

SMILES string

Cl.NCCC[C@H](N)C(O)=O

InChI

1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m0./s1

InChI key

GGTYBZJRPHEQDG-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Product of arginine degradation by arginase

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sherry Dadsetan et al.
Biochemical pharmacology, 85(1), 115-123 (2012-10-30)
Combined administration of ornithine and phenylacetate (OP) is proposed as a novel treatment of hyperammonemia and hepatic encephalopathy. Ornithine is believed to increase ammonia fixation into glutamine in muscle tissue and glutamine is subsequently thought to react with phenylacetate forming
Tom Rimaux et al.
Research in microbiology, 164(3), 216-225 (2012-11-28)
Arginine conversion through the arginine deiminase (ADI) pathway is a common metabolic trait of Lactobacillus sakei which is ascribed to an arc operon and which inquisitively involves citrulline excretion and re-uptake. The aim of this study was to verify whether
Charles D Mills
Critical reviews in immunology, 32(6), 463-488 (2013-02-23)
The purpose of immunology is simple. Cure or prevent disease. M1/M2 is useful because it is simple. M1/M2 describes the two major and opposing activities of macrophages. M1 activity inhibits cell proliferation and causes tissue damage while M2 activity promotes
Irene Tsiapa et al.
Nuclear medicine and biology, 40(2), 262-272 (2012-12-15)
Radiolabeled RGD peptides that specifically target integrin α(ν)β(3) have great potential in early tumor detection through noninvasive monitoring of tumor angiogenesis. Based on previous findings of our group on radiopeptides containing positively charged aminoacids, we developed a new cyclic cRGDfK
Nicole Berthold et al.
Antimicrobial agents and chemotherapy, 57(1), 402-409 (2012-11-02)
Proline-rich antimicrobial peptides (PrAMPs) from insects and mammals have recently been evaluated for their pharmaceutical potential in treating systemic bacterial infections. Besides the native peptides, several shortened, modified, or even artificial sequences were highly effective in different murine infection models.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service