75403
Crystallization Cryo Kit for Proteins
Synonym(s):
Protein Cryo Crystallography Kit
About This Item
Recommended Products
storage temp.
2-8°C
Application
Features and Benefits
- Comprehensive, efficient screening
- Stringent purity standards
- Prefabricated reagents, flow charts, and observation sheets
- Low incidence of undesirable solubility effects
Components
Principle
Kit Components Only
- Ammonium sulfate 0.17 M – Polyethylene glycol 8000 25.5% (w/v) – Glycerol 15% solution
Kit Components Also Available Separately
- 71591Imidazole pH 6.5; 0.07 M - Sodium acetate 0.7 M - Glycerol 30% solutionSDS
- Sodium citrate tribasic pH 5.6; 0.07 M – Ammonium phosphate monobasic 0.7 M – Glycerol 30% (v/v) solution
- 72872Magnesium chloride 0.18 M – HEPES-Na pH 7.5; 0.09M – 2-Propanol 27% (w/v) – Glycerol 10% solutionSDS
- 74613Ammonium acetate 0.16M – TRIS hydrochloride pH 8.5; 0.08M – 2-Propanol 24% (v/v) – Glycerol 20% (v/v) solutionSDS
- Magnesium chloride 0.2M – HEPES Na pH 7.5; 0.1M – Polyethylene glycol 400 30% (w/v) solution
- HEPES Na pH 7.5; 0.065M – Potassium sodium tartrate 0.52M – Glycerol 35% (v/v) solution
- Ammonium acetate 0.17M – Sodium acetate pH 4.6; 0.085M – Polyethylene glycol 4000 25.5% (w/v) – Glycerol 15% (v/v) solution
- Magnesium acetate 0.2M – Sodium cacodylate pH 6.5; 0.1M – 2-Methyl-2,4-pentanediol 30% (v/v) solution
- Sodium citrate tribasic 0.2M – TRIS hydrochloride pH 8.5; 0.1M – Polyethylene glycol 400 30% (w/v) solution
- 80074Ammonium sulfate 0.17M – Sodium cacodylate pH 6.5; 0.085M – Polyethylene glycol 8000 25.5% (w/v) – Glycerol 15% (v/v) solutionSDS
- Lithium sulfate 0.17 M – TRIS hydrochloride pH 8.5; 0.085M – Polyethylen glycol 4000 25.5% (w/v) – Glycerol 15% (v/v) solution
- Magnesium acetate 0.16 M – Sodium cacodylate pH 6.5; 0.08M – Polyethylene glycol 8000 16% (w/v) – Glycerol 20% solution
- 80384Sodium citrate tribasic 0.14M, HEPES Na pH 7.5; 0.07M, 2-Propanol 14% (v/v), Glycerol 30% solutionSDS
- 80387Sodium acetate 0.17M – Sodium cacodylate pH 6.5; 0.085M – Polyethylene glycol 8000 25.5% (w/v) – Glycerol 15% (v/v) solutionSDS
- Calcium chloride 0.19 M – HEPES-Na pH 7.5; 0.095M – Polyethylene glycol 400; 26.6% (w/v) – Glycerol 5% solution
- HEPES-Na pH 7.5; 0.085M – Polyethylene glycol 400 1.7% (v/v) – Ammonium sulfate 1.7M – Glycerol 15% (v/v) solution
- HEPES-Na pH 7.5; 0.075M – Sodium phosphate monobasic 0.6M – Potassium phosphate monobasic 0.6M – Glycerol 25% (v/v) solution
- Lithium sulfate 0.4 M – Polyethylene glycol 8000 12% (w/v) – Glycerol 20% (v/v) solution
- 81892Sodium formate 3.6 M – Glycerol 10% solutionSDS
- Sodium acetate pH 4.6; 0.07M – 1.4 M Sodium formate – Glycerol 30% (v/v) solution
- HEPES-Na pH 7.5; 0.085M – 2-Propanol 8.5% (v/v) – Polyethylene glycol 4000 17% (w/v) – Glycerol 15% (v/v) solution
- 83196Sodium acetate pH 4.6; 0.08 M – Ammonium sulfate 1.6 M – Glycerol 20% (v/v) solutionSDS
- Potassium phosphate monobasic 0.04M – Polyethylene glycol 8000 16% (w/v) – Glycerol 20% (v/v) solution
- TRIS hydrochloride pH 8.5; 0.08 M – Ammonium phosphate monobasic 1.6 M – Glycerol 20% (v/v) solution
- 89252Ammonium sulfate 1.5 M – Glycerol 25% solutionSDS
- HEPES-Na pH 7.5; 0.09M – Sodium citrate tribasic 1.26M – Glycerol 10% solution
- Polyethylene glycol 1500 24% (w/v) – Glycerol 20% (v/v) solution
- 91638Magnesium formate 0.1M – Glycerol 50% (v/v) solutionSDS
- Calcium chloride 0.02 M – Sodium acetate 0.1M – 2-Methyl-2,4-pentanediol 30% (v/v) solution
- Sodium citrate tribasic 0.2M – HEPES-Na pH 7.5; 0.1M – 2-Methyl-2,4-pentanediol 30% (v/v) solution
- TRIS hydrochloride pH 8.5; 0.075 M – Ammonium sulfate 1.5 M – Glycerol 25% (v/v) solution
- Ammonium phosphate monobasic 0.26 M – Glycerol 35% (v/v) solution
- 96864Potassium sodium tartrate 0.26 M – Glycerol 35% (v/v) solutionSDS
- Calcium chloride 0.14M – Sodium acetate pH 4.6; 0.07M – 2-Propanol 14% (v/v) – Glycerol 30% (v/v) solution
- Sodium cacodylate pH 6.5; 0.07M – Sodium acetate 0.98M – Glycerol 30% (v/v) solution
- Ammonium sulfate 0.16 M – Sodium acetate pH 4.6; 0.08M – Polyethylene glycol 4000 20% (w/v) – Glycerol 20% (v/v) solution
- 80084HEPES Na pH 7.5; 0.075M – Lithium sulfate 1.125M – Glycerol 25% (v/v) solutionSDS
signalword
Danger
Hazard Classifications
Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service