This product contains 500mg of Orange G dye and 15mg of Phosphotungstic acid in 100ml of Ethanol.
75400
Orange G solution
alcoholic, for the colorimetric method according to Papanicolaou
Synonym(s):
OG-6
About This Item
Recommended Products
form
liquid
density
0.80 g/mL at 20 °C
λmax
0.785-0.814 at 493-503 nm in ethanol
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
[Na+].[Na+].Oc1ccc2cc(cc(c2c1\N=N\c3ccccc3)S([O-])(=O)=O)S([O-])(=O)=O
InChI
1S/C16H12N2O7S2.2Na/c19-13-7-6-10-8-12(26(20,21)22)9-14(27(23,24)25)15(10)16(13)18-17-11-4-2-1-3-5-11;;/h1-9,19H,(H,20,21,22)(H,23,24,25);;/q;2*+1/p-2/b18-17+;;
InChI key
HSXUHWZMNJHFRV-QIKYXUGXSA-L
Looking for similar products? Visit Product Comparison Guide
Caution
Other Notes
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
57.2 °F - closed cup
flash_point_c
14.0 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Hallo, wie viel prozentig ist diese Lösung denn? Lieben Gruß, Louis
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service