Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75378

Sigma-Aldrich

(R)-γ-Caprolactone

≥98.5% (GC)

Synonym(s):

(+)-γ-Hexalactone, (5R)-5-Ethyldihydro-2(3H)-furanone, (5R)-5-Ethyloxolan-2-one, (R)-γ-Ethyl-γ-butyrolactone, (R)-4-Hexanolide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
Beilstein/REAXYS Number:
3587788
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

assay

≥98.5% (GC)

form

liquid

optical activity

[α]20/D +53±1°, c = 1 in methanol

optical purity

enantiomeric excess: ≥95.0% (GC)

storage temp.

2-8°C

SMILES string

CC[C@@H]1CCC(O1)=O

InChI

1S/C6H10O2/c1-2-5-3-4-6(7)8-5/h5H,2-4H2,1H3/t5-/m1/s1

InChI key

JBFHTYHTHYHCDJ-RXMQYKEDSA-N

Biochem/physiol Actions

Metabolite produced by plants, as a component of aroma flavours and fragrances and aromas of fruits and vegetablest[1]. Component of the pheromone secreted by the female dermestid beetle Trogoderma glabrum and Trogoderma granarium[2]. gamma-Caprolactone is occasionally found as a volatile component of human urine and has been found in the polar fraction of human blood. Biological fluids such as blood and urine have been shown to contain a large number of components apparently present in all individuals. It is a promotor of the growth of biocontrol bacteria and a metabolite of a new assimilative pathway.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sex pheromone of the female dermestid beetle Trogoderma glabrum.
Yarger, R. G., et al.
Journal of Chemical Ecology, 1, 323-334 (1975)
Stereoisomeric flavor compounds: structure and properties of gamma-lactone enantiomers.
Mosandl, A. and Gunther, C.
Journal of Agricultural and Food Chemistry, 37, 413-418 (1989)
Carlo Bicchi et al.
Journal of chromatography. A, 1212(1-2), 114-123 (2008-10-31)
The analysis of complex real-world samples of vegetable origin requires rapid and accurate routine methods, enabling laboratories to increase sample throughput and productivity while reducing analysis costs. This study examines shortening enantioselective-GC (ES-GC) analysis time following the approaches used in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service