Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75166

Sigma-Aldrich

Biotin-B-Phycoerythrin

BioReagent, suitable for gel electrophoresis

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352108
NACRES:
NA.32

product line

BioReagent

form

liquid

mol wt

Mr ~240000

composition

BSA, 0.1%
pigment, 2 mg/mL
sodium azide, 0.05%
sodium chloride, 150 mM
sodium phosphate pH7.2, 100 mM
sucrose, 50 mM

technique(s)

electrophoresis: suitable

fluorescence

λex 542 nm; λem 576 nm in 0.1 M phosphate pH 7.0

suitability

suitable for gel electrophoresis

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Caution

Do not freeze, store at 4°C in the dark.

Analysis Note

A545/A280 ≥ 3.5; A565/A545 ≤ 1.0; A620/A545 ≤ 0.02

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service