Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74993

Sigma-Aldrich

D-Aminoacylase recombinant from Escherichia coli

≥250 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

crystals
powder

specific activity

≥250 U/mg

storage temp.

−20°C

Biochem/physiol Actions

D -aminoacylase catalyzes the zinc-assisted hydrolysis of N-acyl- D -amino acids to produce D -amino acids.[1]

Unit Definition

One unit corresponds to the amount of enzyme which hydrolyzes 1 μmol N-acetyl-DL-methionine per minute at pH 8.0 and 37°C.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shwu-Huey Liaw et al.
The Journal of biological chemistry, 278(7), 4957-4962 (2002-11-28)
D-Aminoacylase is an attractive candidate for commercial production of D-amino acids through its catalysis in the hydrolysis of N-acyl-D-amino acids. We report here the first D-aminoacylase crystal structure from A. faecalis at 1.5-A resolution. The protein comprises a small beta-barrel

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service