Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74718

Sigma-Aldrich

Octaethylene glycol monooctadecyl ether

BioXtra, ≥98.0% (TLC)

Synonym(s):

C18E8, Octadecyl octaethylene glycol ether, Octadecyloctaglycol, Polyoxyethylene (8) stearyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17(OCH2CH2)8OH
CAS Number:
Molecular Weight:
622.91
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

product line

BioXtra

assay

≥98.0% (TLC)

mol wt

622.91 g/mol

technique(s)

protein quantification: suitable

mp

47-49 °C

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C34H70O9/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-36-21-23-38-25-27-40-29-31-42-33-34-43-32-30-41-28-26-39-24-22-37-20-18-35/h35H,2-34H2,1H3

InChI key

WPXCYCTVMORDCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service