Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74432

Sigma-Aldrich

Nonylphenyl-polyethyleneglycol acetate

suitable for histology

Synonym(s):

Polyethylene glycol nonylphenyl ether acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H19C9C6H4(OCH2CH2)nOCOCH3
CAS Number:
MDL number:
UNSPSC Code:
12161900

description

non-ionic

form

liquid

impurities

≤0.006% peroxides (as H2O2)
≤1% acetic anhydride

acid value

≤1.5

ester number

82‑90

application(s)

histology

Application

Plasticizer for embedding bone-containing tissue for histochemistry[1][2]

related product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Preparative prerequisites in clinical histology of human bone marrow. 1. Methodical studies on embedding in acrylates of lipid-rich tissue material].
R Burkhardt
Blut, 13(6), 337-357 (1966-09-01)
Leukokinetic studies. I. A method for labeling leukocytes with diisopropyl-fluorophosphate (DFP32).
J W ATHENS et al.
Blood, 14(4), 303-333 (1959-04-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service