Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74414

Sigma-Aldrich

(S)-3-Methyl-2-oxovaleric acid lithium salt hydrate

≥95.0% (TLC)

Synonym(s):

(S)-3-Methyl-2-oxopentanoic acid, (S)-Ketoisoleucine lithium salt, Lithium (S)-3-methyl-2-oxovalerate, S-KMV-Li

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9LiO3 · xH2O
CAS Number:
Molecular Weight:
136.07 (anhydrous basis)
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

Product Name

(S)-3-Methyl-2-oxovaleric acid lithium salt hydrate, ≥95.0% (TLC)

assay

≥95.0% (TLC)

form

powder or crystals

optical activity

[α]/D +21.5±2°, c = 0.1 in 0.1 M HCl

color

white to off-white

storage temp.

2-8°C

SMILES string

[Li+].O.CC[C@H](C)C(=O)C([O-])=O

InChI

1S/C6H10O3.Li.H2O/c1-3-4(2)5(7)6(8)9;;/h4H,3H2,1-2H3,(H,8,9);;1H2/q;+1;/p-1/t4-;;/m0../s1

InChI key

PTRNELNRMVEQEC-FHNDMYTFSA-M

Biochem/physiol Actions

Metabolite in valine, leucine and isoleucine biosynthesis and in glucosinolate biosynthesis. S-KMV is found in human plasma, the S-KMV:R-KMV ratio is used for diagnosis of diabetes mellitus and maple syrup urine disease.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of R- and S-3-methyl-2-oxopentanoate enantiomers in human plasma: suitable method for label enrichment analysis.
Schadewaldt, P., et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 682, 209-218 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service