Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74393

Sigma-Aldrich

N-(Carbonyl-methoxypolyethylenglycol 2000)-1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine sodium salt

≥98.0% (TLC)

Synonym(s):

1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine, Carbonyl-methoxypolyethylenglycol 2000 labeled, MPEG-2000-DMPE Na

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.85

Quality Level

assay

≥98.0% (TLC)

form

powder

functional group

PEG
phospholipid

lipid type

phospholipids

shipped in

ambient

storage temp.

room temp

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service