Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74359

Sigma-Aldrich

N-[(RS)-3-Hydroxybutyryl]-L-homoserine lactone

≥96% (HPLC)

Synonym(s):

3-Hydroxy-C4-HSL, N-(3-Hydroxybutanoyl)-L-homoserine lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13NO4
CAS Number:
Molecular Weight:
187.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥96% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

CC(O)CC(=O)N[C@H]1CCOC1=O

InChI

1S/C8H13NO4/c1-5(10)4-7(11)9-6-2-3-13-8(6)12/h5-6,10H,2-4H2,1H3,(H,9,11)/t5?,6-/m0/s1

InChI key

FIXDIFPJOFIIEC-GDVGLLTNSA-N

Biochem/physiol Actions

N-[(RS)-3-Hydroxybutyryl]-L-homoserine lactone is a member of N-acyl-homoserine lactone family. N-Acylhomoserine lactones (AHL) regulate gene expression in gram-negative bacteria, such as Echerichia and Salmonella, and are involved in quorum sensing, cell to cell communication among bacteria; for reviews see[1][2][3][4][5]. Bacterial intercellular communication has become a target for the development of new anti-virulence drugs[3], and a research focus for the prevention of biofilm formation[6][7][8].
Quorum-sensing signal generation

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B L Hanzelka et al.
Journal of bacteriology, 181(18), 5766-5770 (1999-09-11)
Acylhomoserine lactones, which serve as quorum-sensing signals in gram-negative bacteria, are produced by members of the LuxI family of synthases. LuxI is a Vibrio fischeri enzyme that catalyzes the synthesis of N-(3-oxohexanoyl)-L-homoserine lactone from an acyl-acyl carrier protein and S-adenosylmethionine.
Sathish Rajamani et al.
Journal of microbiological methods, 84(2), 189-193 (2010-12-07)
Many bacteria produce and secrete proteases during host invasion and pathogenesis. Vibrio harveyi, an opportunistic pathogen of shrimp, is known to use a two-component quorum sensing (QS) mechanism for coordination of gene expression including protease secretion at high population densities.
Bonnie L Bassler
Cell, 109(4), 421-424 (2002-06-28)
In a process called quorum sensing, groups of bacteria communicate with one another to coordinate their behavior and function like a multicellular organism. A diverse array of secreted chemical signal molecules and signal detection apparatuses facilitate highly productive intra- and
M B Miller et al.
Annual review of microbiology, 55, 165-199 (2001-09-07)
Quorum sensing is the regulation of gene expression in response to fluctuations in cell-population density. Quorum sensing bacteria produce and release chemical signal molecules called autoinducers that increase in concentration as a function of cell density. The detection of a
Jie Zhu et al.
Current opinion in pharmacology, 13(5), 688-698 (2013-07-24)
With the emergence of microbial pathogens increasingly resistant against commonly used antibiotics, new treatment strategies are desperately needed. Bacterial quorum sensing has attracted a lot of attention over the last decade as a potential new target for antimicrobial therapy. Interference

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service