Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74171

Sigma-Aldrich

N-Fmoc Rhodamine 110

suitable for fluorescence

Synonym(s):

3-Amino-9-(2-carboxyphenyl)-6-[[(9H-fluoren-9-ylmethoxy)carbonyl]amino]-xanthylium chloride (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H25ClN2O5
CAS Number:
Molecular Weight:
589.04
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

form

powder

fluorescence

λex 492 nm; λem 521.0 nm±10 nm in ethanol

suitability

suitable for fluorescence

functional group

Fmoc

storage temp.

−20°C

SMILES string

NC1=CC=C(C(C2=CC=CC=C2C(O)=O)=C(C=CC(NC(OCC3C(C=CC=C4)=C4C5=C3C=CC=C5)=O)=C6)C6=[O+]7)C7=C1.[Cl-]

InChI

1S/C35H24N2O5.ClH/c36-20-13-15-28-31(17-20)42-32-18-21(14-16-29(32)33(28)26-11-5-6-12-27(26)34(38)39)37-35(40)41-19-30-24-9-3-1-7-22(24)23-8-2-4-10-25(23)30;/h1-18,30H,19,36H2,(H-,37,38,39,40);1H

InChI key

RBQWVYBTQCULOF-UHFFFAOYSA-N

General description

Mono-protected rhodamine 110 derivative useful for the synthesis of heterofunctional R110 derivatives, including peptidase substrates.

Note :
EX = 492nm for free R110 (e=88K)

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service