Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74006

Sigma-Aldrich

Isopyridoxal hydrochloride

≥98.5% (HPLC)

Synonym(s):

5-Hydroxy-4-(hydroxymethyl)-6-methyl-3-pyridinecarboxaldehyde hydrochloride, 5-Hydroxy-4-(hydroxymethyl)-6-methylnicotinaldehyde hydrochloride, 5-Hydroxy-4-(hydroxymethyl)-6-methylpyridine-3-carbaldehyde hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO3· HCl
CAS Number:
Molecular Weight:
203.62
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.79

assay

≥98.5% (HPLC)
98.5-101.5% (T)

form

powder

color

white

suitability

corresponds for identity (IR)

application(s)

cell analysis

storage temp.

2-8°C

SMILES string

CC1=C(O)C(CO)=C(C([H])=O)C=N1.Cl

InChI

1S/C8H9NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2-3,11-12H,4H2,1H3;1H

InChI key

IJRYEEAKAHJPTM-UHFFFAOYSA-N

Biochem/physiol Actions

Isopyridoxal is an active vitamer of the B6 complex in humans[1]; enzyme substrate for isopyridoxal dehydrogenase[2]; metabolite standard in the determination of all seven vitamin B6-related compounds[3][1]; and metabolite in a pyridoxine degradation pathway by soil bacteria[4].

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bacterial oxidation of vitamin B6. I. Isopyridoxal and 5-pyridoxic acid.
V W RODWELL et al.
The Journal of biological chemistry, 233(6), 1548-1554 (1958-12-01)
Baiqiang Yuan et al.
FEMS microbiology letters, 234(2), 225-230 (2004-05-12)
The gene product of mll6785 of a nitrogen-fixing symbiotic bacterium Mesorhizobium loti MAFF303099 was identified as pyridoxine 4-oxidase, the first enzyme in the vitamin B6-degradation pathway. The gene was cloned and ligated into pET-21a+. Escherichia coli BL21(DE3) was co-transformed with
Y C Lee et al.
The Journal of biological chemistry, 261(32), 15106-15111 (1986-11-15)
Two NAD+-dependent, highly specific pyridine-5-aldehyde dehydrogenases, 5-formyl-3-hydroxy-2-methylpyridine-4-carboxylic-acid (Compound 1) dehydrogenase and isopyridoxal dehydrogenase, were purified to homogeneity from Pseudomonas MA-1 and Arthrobacter Cr-7, respectively. Both enzymes are induced in response to growth of the organisms on pyridoxine and catalyze steps
P Edwards et al.
Clinica chimica acta; international journal of clinical chemistry, 190(1-2), 67-80 (1990-09-01)
The effects of increased intake of pyridoxine hydrochloride on plasma vitamin B6 metabolism within therapeutic limits (up to 800 mg/day) were investigated. Maximum plasma concentrations of pyridoxal phosphate were attained at relatively low intakes of pyridoxine hydrochloride. Two metabolism thought
C J Argoudelis
Journal of chromatography. A, 790(1-2), 83-91 (1998-01-22)
A simple, sensitive, isocratic high-performance liquid chromatographic method has been developed for the separation of all seven vitamin B6-related compounds. The separation is accomplished using an ODS column and a mobile phase of 0.15 M sodium dihydrogenphosphate, adjusted to pH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service