Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73933

Sigma-Aldrich

Sodium citrate tribasic pH 5.6; 0.1M – 2-Propanol 20% – PEG 4000 20% solution

Synonym(s):

Crystallization solution 40/Kit-No 82009

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

liquid

pH

5.6±0.2 ( in neat)

density

1.02 g/mL at 20 °C

storage temp.

2-8°C

pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

75.2 °F - closed cup

flash_point_c

24.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service