Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73806

Sigma-Aldrich

Calcium chloride 0.14M – Sodium acetate pH 4.6; 0.07M – 2-Propanol 14% (v/v) – Glycerol 30% (v/v) solution

Synonym(s):

Crystallization solution 24/Kit No 75403

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available. Contact Technical Service

form

solution

pH

4.6±0.2 ( in neat)

density

1.08 g/mL at 20 °C

storage temp.

2-8°C

pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

80.6 °F

flash_point_c

27 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service