Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73498

Sigma-Aldrich

cis-Inositol

≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein/REAXYS Number:
2042263
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

assay

≥98.0% (TLC)

storage temp.

room temp

SMILES string

O[C@@H]1[C@H](O)[C@H](O)[C@H](O)[C@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2+,3-,4+,5-,6+

InChI key

CDAISMWEOUEBRE-JMVOWJSSSA-N

Application

cis-Inositol was used to improve the intensity and desorption of samples during field desorption mass spectrometry analysis.

Biochem/physiol Actions

Cis-inositol is one of the six possible steroisomer of inositol, a cyclohexane polyol. The cis-inositol isomer readily forms strong complexes with cations and oxyacid anions.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancement of field desorption mass spectra of alkali metal salts by polyhydroxyl additives.
Wood, G.
Biological Mass Spectrometry, 9, 72-73 (1982)
Ileana Andreea Ratiu et al.
Molecules (Basel, Switzerland), 25(1) (2019-12-22)
Honey is a natural sweetener, with an osmotic effect on microorganisms due to the increased sugar content and low amount of water. Cyclitols are minor constituents of honey. They play a defensive role in plants against unfavorable environmental conditions. Honey's
Sándor L Bekö et al.
IUCrJ, 1(Pt 1), 61-73 (2014-07-31)
Inositol, 1,2,3,4,5,6-hexahydroxycyclohexane, exists in nine stereoisomers with different crystal structures and melting points. In a previous paper on the relationship between the melting points of the inositols and the hydrogen-bonding patterns in their crystal structures [Simperler et al. (2006 ▶).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service