Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72721

Sigma-Aldrich

AZCL-Pachyman

substrate for laminarinase

Synonym(s):

Azurine-cross-linked pachyman

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

powder

storage temp.

2-8°C

Application

This enzyme substrate is prepared by dyeing and cross-linking highly purified pachyman, ß-1,3-D-glucan isolated from a Basidiomycetes sp. This enzyme substrate, which hydrates but is insoluble in water, can be used for measuring hydrolysis by endo-1,3-ß-glucanase. Enzyme activities can be measured by the rate of release of the water-soluble dyed fragments (increase in absorbance at 590 nm).
suitable as substrate for laminarinase

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service