Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72572

Sigma-Aldrich

Fluorescent red NIR 680

suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H46N2O6S
Molecular Weight:
634.83
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

form

powder

Quality Level

solubility

DMSO: soluble

fluorescence

λex 680 nm; λem ~715 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

General description

Fluorescent red 680 is a fluorescent dye for the near infrared area. Extremely long wavelength fluorescence is especially well suited for applications where sample autofluorescence might be critical or penetration of tissue or other matrix is required.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service