Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72172

Sigma-Aldrich

Fluorescent IEF-Marker pI 3.0 solution

Synonym(s):

IEF-Marker fluorescent pI 3.0

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105336
NACRES:
NA.26

form

liquid

mol wt

Mw 215.63 g/mol

concentration

2 mg/mL in 5 mM HCl, 50% 2-propanol

fluorescence

λex 360 nm; λem 445 nm in 0.1 M citrate pH 3.0

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

filtered through a 0.45 μm membrane filter

Application

Suitable as IEF-Marker for HPCE and for IEF-gels

Reconstitution

For use in HPCE the marker should be diluted 1:100 in a suitable ampholyte solution (e.g. 2% ampholyte 3-10), UV-detection at 280 nm (20 °C). For use in IEF-gels the solution can be loaded directly onto the gel (max. 1μl per lane).

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

53.6 °F

flash_point_c

12.0 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Information on Isoelectric Focusing including what it is and how it is used. In order to ensure the high performance of analysis, isoelectric point (pI) standards are needed.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service