Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69299

Sigma-Aldrich

TNP-GTP triethylammonium salt

≥95.0% (HPLC)

Synonym(s):

2′,3′-O-Trinitrophenylguanosine 5′-triphosphate triethylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N8O20P3 · xC6H15N
CAS Number:
Molecular Weight:
734.27 (free acid basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

assay

≥95.0% (HPLC)

form

solid

pH

7.0-8.0

storage temp.

−20°C

SMILES string

OP(OP(O)(OP(O)(OC[C@@H]1[C@@H](OC2(C([N+]([O-])=O)=CC(C=C2[N+]([O-])=O)=[N+]([O-])O)O3)C3[C@H](N(C=N4)C5=C4C(NC(N)=N5)=O)O1)=O)=O)(O)=O.CCN(CC)CC

InChI

1S/C16H17N8O20P3.C6H15N/c17-15-19-12-9(13(25)20-15)18-4-21(12)14-11-10(6(40-14)3-39-46(35,36)44-47(37,38)43-45(32,33)34)41-16(42-11)7(23(28)29)1-5(22(26)27)2-8(16)24(30)31;1-4-7(5-2)6-3/h1-2,4,6,10-11,14H,3H2,(H,26,27)(H,35,36)(H,37,38)(H2,32,33,34)(H3,17,19,20,25);4-6H2,1-3H3/t6-,10-,11?,14-;/m1./s1

InChI key

MSJYAQKTVLFNEE-LGQBSHKBSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service