Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69296

Sigma-Aldrich

Fluorescent red 636 reactive

suitable for fluorescence, ≥70% (coupling to amines)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H48N3NaO11S2
Molecular Weight:
833.94
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥70% (coupling to amines)

form

powder

fluorescence

λex 636 nm; λem 665 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[Na+].CCN(CC)c1ccc2c(OC(=C\C2=C/C=C/C3=[N+](CCCS([O-])(=O)=O)c4ccc(cc4C3(C)CCCC(=O)ON5C(=O)CCC5=O)S([O-])(=O)=O)C(C)(C)C)c1

InChI

1S/C40H49N3O11S2.Na/c1-7-41(8-2)28-15-17-30-27(24-35(39(3,4)5)53-33(30)25-28)12-9-13-34-40(6,21-10-14-38(46)54-43-36(44)19-20-37(43)45)31-26-29(56(50,51)52)16-18-32(31)42(34)22-11-23-55(47,48)49;/h9,12-13,15-18,24-26H,7-8,10-11,14,19-23H2,1-6H3,(H-,47,48,49,50,51,52);/q;+1/p-1

InChI key

FLVFZXRJCNTEDK-UHFFFAOYSA-M

General description

Fluorescent Red 636 reactive is a fluorescent label especially well suited for excitation by He-Ne-lasers (at 633 nm). Fluorescent red 636 reactive shows strong fluorescence (molar absorption 190 000 M-1cm-1) and good solubility in water.

application

This fluorescent label has a very high molar absorption (190,000).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service