Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69104

Sigma-Aldrich

Propargylamino-dCTP-Cy®5 triethylammonium salt solution

≥95.0% (HPLC)

Synonym(s):

5-Propargylamino-2′-deoxycytidine 5′-triphosphate labeled with Cy®5 triethylammonium salt, Cy5-AP3-dCTP, Cy®5 5-(3-Aminopropargyl)-2′-deoxycytidine 5′-triphosphate triethylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H57N6O20P3S2 · xC6H15N
Molecular Weight:
1159.01 (free acid basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

assay

≥95.0% (HPLC)

form

liquid

pH

7.0-8.0

storage temp.

−20°C

SMILES string

OP(OP(O)(OP(O)(OC[C@@H]1[C@@H](O)C[C@H](N2C(N=C(N)C(C#CCNC(CCCCCN(C3=C(C/4(C)C)C=C(S(=O)(O)=O)C=C3)C4=C\C=C\C=C\C5=[N+](CC)C6=CC=C(S(=O)([O-])=O)C=C6C5(C)C)=O)=C2)=O)O1)=O)=O)(O)=O.CCN(CC)CC

InChI

1S/C45H57N6O20P3S2.C6H15N/c1-6-49-34-20-18-30(75(62,63)64)24-32(34)44(2,3)38(49)15-9-7-10-16-39-45(4,5)33-25-31(76(65,66)67)19-21-35(33)50(39)23-12-8-11-17-40(53)47-22-13-14-29-27-51(43(54)48-42(29)46)41-26-36(52)37(69-41)28-68-73(58,59)71-74(60,61)70-72(55,56)57;1-4-7(5-2)6-3/h7,9-10,15-16,18-21,24-25,27,36-37,41,52H,6,8,11-12,17,22-23,26,28H2,1-5H3,(H8-,46,47,48,53,54,55,56,57,58,59,60,61,62,63,64,65,66,67);4-6H2,1-3H3/t36-,37-,41-;/m1./s1

InChI key

QIQHTLLIYIAFGQ-ATBZKAILSA-N

Legal Information

Cy is a registered trademark of Cytiva

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel Castañeda-Zárate et al.
Current biology : CB, 31(1), 238-246 (2020-11-07)
During the evolutionary history of flowering plants, transitions between pollinator groups (pollinator shifts) have been frequent,1 and contributed to the spectacular radiation of angiosperms.2 Although the evolution of floral traits during pollinator shifts has been studied in real time under

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service