Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69061

Sigma-Aldrich

Dihydropinosylvin monomethyl ether

≥97.0% (HPLC)

Synonym(s):

3-Hydroxy-5-methoxybibenzyl, 3-Methoxy-5-(2-phenylethyl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O2
CAS Number:
Molecular Weight:
228.29
Beilstein/REAXYS Number:
1971738
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥97.0% (HPLC)

form

powder

mp

49-53 °C

SMILES string

COc1cc(O)cc(CCc2ccccc2)c1

InChI

1S/C15H16O2/c1-17-15-10-13(9-14(16)11-15)8-7-12-5-3-2-4-6-12/h2-6,9-11,16H,7-8H2,1H3

InChI key

HPEFWCAKFRCLBD-UHFFFAOYSA-N

Application

Dihydropinosylvin monomethyl ether may be used in studies of stilbenoid chemistry and function.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Fliegmann et al.
Plant molecular biology, 18(3), 489-503 (1992-02-01)
Chalcone synthase (CHS) and stilbene synthase (STS) are closely related polyketide synthases which are key enzymes in the biosynthesis of flavonoids and stilbenes. Scots pine (Pinus sylvestris) is an interesting plant for a direct comparison of the enzymes. It not
Michael Adams et al.
Journal of natural products, 68(1), 83-85 (2005-02-01)
Fifteen stilbenoids and two alkaloids from Stemona collinsae, S. tuberosa, and S. peirrei were tested alongside the commercially available stilbenoids resveratrol and pinosylvin for inhibition of leukotriene formation in an ex vivo test system based on activated human neutrophilic granulocytes.
L E Lindberg et al.
Journal of industrial microbiology & biotechnology, 31(3), 137-147 (2004-04-28)
Hydrophilic knotwood extracts from 18 wood species were assessed in disc diffusion and liquid culture tests for antibacterial effects against three species of paper mill bacteria. The Pinus sylvestris, P. resinosa, P. contorta, and P. banksiana extracts decreased or inhibited

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service