Recommended Products
form
solid
size
57 mm × 130 mm
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- High porosity of the strip (approx. 80 % free pore volume)
- Isotropic, integral matrix for ideal flow conditions
- Minimal diffusion effect and low adsorption capacity
- Perfect transparency because of the pore system
- Ideal for the non-toxic ATX reagent system
- Virtually unlimited stability at room temperature
- Two identical sides, no need to search for the "right" side
These strips can also be used for high-voltage electrophoresis, chromatography, and immunodiffusion.
Storage Class
13 - Non Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service