68653
Ampholyte high resolution
pH 4.0-6.0, ~40% in H2O
Synonym(s):
SinuLyte™
About This Item
Recommended Products
form
liquid
Quality Level
concentration
~40% in H2O
technique(s)
electrophoresis: suitable
pH
4.0-6.0
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
- High buffer capacity: Compared to established ampholytes they offer buffering capacities 1.5-2.5 times higher (up to pH 8, equal above pH 8)
- Good, even conductivity
- Linear and reproducible pH gradient
- High resolution
Legal Information
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service