Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67858

SAFC

Dy-554 NHS ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H37ClN4O8
Molecular Weight:
677.14
UNSPSC Code:
12352200

form

solid

fluorescence

λex 554 nm; λem 579 nm in 0.1 M phosphate pH 7.0

suitability

complies for coupling to amines
suitable for fluorescence

storage temp.

2-8°C

Application

Dy-554 NHS ester, an amine coupling asymmetric cyanine reagent, is used for fluorescent labeling of amine groups within proteins, amino derivatized oligonucleotide and other amine containing molecules.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jutta Pauli et al.
Bioconjugate chemistry, 22(7), 1298-1308 (2011-05-19)
Aiming at the design of highly brilliant NIR emissive optical probes, e.g., for in vivo near-infrared fluorescence imaging (NIRF), we studied the absorption and fluorescence properties of the asymmetric cyanines Dy678, Dy681, Dy682, and Dy676 conjugated to the model antibody
Franziska M Hamann et al.
Molecular imaging, 10(4), 258-269 (2011-04-28)
To assess the suitability of asymmetric cyanine dyes for in vivo fluoro-optical molecular imaging, a comprehensive study on the influence of the number of negatively charged sulfonate groups governing the hydrophilicity of the DY-67x family of asymmetric cyanines was performed.
Marcia Levitus et al.
Quarterly reviews of biophysics, 44(1), 123-151 (2010-11-27)
The breakthroughs in single molecule spectroscopy of the last decade and the recent advances in super resolution microscopy have boosted the popularity of cyanine dyes in biophysical research. These applications have motivated the investigation of the reactions and relaxation processes
Judith E Berlier et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 51(12), 1699-1712 (2003-11-19)
Amine-reactive N-hydroxysuccinimidyl esters of Alexa Fluor fluorescent dyes with principal absorption maxima at about 555 nm, 633 nm, 647 nm, 660 nm, 680 nm, 700 nm, and 750 nm were conjugated to antibodies and other selected proteins. These conjugates were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service