Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67395

Sigma-Aldrich

Octyl β-D-1-thioglucopyranoside

≥99% (TLC)

Synonym(s):

Octyl thioglucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28O5S
CAS Number:
Molecular Weight:
308.43
Beilstein/REAXYS Number:
4182783
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

assay

≥99% (TLC)

optical activity

[α]/D -52 to -48°, c = 1 in ethanol

mol wt

308.43 g/mol

impurities

≤4% water

CMC

9 mM (20-25°C)

solubility

ethanol: 50 mg/mL, clear, colorless

anion traces

sulfate (SO42-): ≤300 mg/kg

cation traces

Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤10 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

CCCCCCCCS[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C14H28O5S/c1-2-3-4-5-6-7-8-20-14-13(18)12(17)11(16)10(9-15)19-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13-,14+/m1/s1

InChI key

CGVLVOOFCGWBCS-RGDJUOJXSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z C Ti et al.
Journal of biotechnology, 16(3-4), 233-243 (1990-11-01)
Large-scale purification of a Dictyostelium discoideum cell surface glycoprotein, which is anchored in the membrane via a glycosylphosphatidylinositol (GPI) moiety, is described. The purification protocol involved four steps: separation of crude cell membranes by low-speed centrifugation, delipidization of these membranes
Use of n-Octyl-b-D-thioglucopyranoside in the isolation of a bacterial membrane protein: an improved method for immunological applications.
Peula-Garcia, J.M., et al.
Journal of Surface Science and Technology, 22(3-4), 141-157 (2006)
Tony Warne et al.
Protein expression and purification, 65(2), 204-213 (2009-03-20)
Structure determination of G protein-coupled receptors is still in its infancy and many factors affect whether crystals are obtained and whether the diffraction is of sufficient quality for structure determination. We recently solved the structure of a thermostabilised turkey beta
K Takeda et al.
Journal of UOEH, 22(4), 317-324 (2001-01-02)
Mouse liver microsomes treated with octylthioglucoside were examined for iron-stimulated ATPase activity. The activity was about 6 mumol Pi/mg protein/hr under optimal conditions [300 mM KC1, 3 mM MgSO4, 10 mM glutathione(GSH), 100 microM FeCl3, 3 mM ATP and 50
Thulasi Warrier et al.
Antimicrobial agents and chemotherapy, 56(4), 1735-1743 (2012-02-01)
The antigen 85 (Ag85) protein family, consisting of Ag85A, -B, and -C, is vital for Mycobacterium tuberculosis due to its role in cell envelope biogenesis. The mycoloyl transferase activity of these proteins generates trehalose dimycolate (TDM), an envelope lipid essential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service