Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67254

Sigma-Aldrich

TFP-PEO-Biotin

Synonym(s):

N-[1-(Biotinylamido)-4,7,10-trioxatridec-13-yl]succinamic acid 2,3,5,6-tetrafluorophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H42F4N4O8S
CAS Number:
Molecular Weight:
694.74
MDL number:
UNSPSC Code:
12352200

form

powder

color

light yellow

application(s)

peptide synthesis

storage temp.

2-8°C

InChI

1S/C30H42F4N4O8S/c31-19-17-20(32)27(34)29(26(19)33)46-25(41)8-7-24(40)36-10-4-12-44-14-16-45-15-13-43-11-3-9-35-23(39)6-2-1-5-22-28-21(18-47-22)37-30(42)38-28/h17,21-22,28H,1-16,18H2,(H,35,39)(H,36,40)(H2,37,38,42)/t21-,22-,28-/m0/s1

InChI key

VIBYBHZETWLVJE-VPYPWEPUSA-N

Application

N-[1-(Biotinylamido)-4,7,10-trioxatridec-13-yl]succinamic acid 2,3,5,6-tetrafluorophenyl ester (TFG-PEO-Biotin) is a reactive biotinylated label with hydrophilic PEO (polyethylene oxide) spacer arm (32.6 Å) used in a solid-phase synthesis of peptide substrate [1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E K Dolence et al.
Bioconjugate chemistry, 12(1), 35-43 (2001-02-15)
Eukaryotic proteins with carboxyl-terminal Ca(1)a(2) motifs undergo three posttranslational processing reactions--prenylation, endoproteolysis, and carboxymethylation. Two genes in yeast encoding Ca(1)a(2)X endoproteases, AFC1 and RCE1, have been identified. Rce1p is solely responsible for proteolysis of yeast Ras proteins. When proteolysis is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service