Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67198

Sigma-Aldrich

Cacodylate pH 6.0; 0.05 M − Magnesium chloride 15 mM − Spermine 3.0 mM − PEG 400 10% solution

Synonym(s):

Crystallization solution 21/Kit-No 80701

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

pH

6.0±0.2 ( in neat)

storage temp.

2-8°C

pictograms

Health hazardEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service