66950
Methyl-β-D-glucopyranoside hemihydrate
≥99.0% (sum of enantiomers, HPLC)
Synonym(s):
β-Methyl-D-glucoside hemihydrate
About This Item
Recommended Products
assay
≥99.0% (sum of enantiomers, HPLC)
optical activity
[α]20/D −33±1°, c = 10% in H2O
mp
107-111 °C
solubility
H2O: 0.1 g/mL, clear, colorless
SMILES string
CO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
InChI
1S/C7H14O6/c1-12-7-6(11)5(10)4(9)3(2-8)13-7/h3-11H,2H2,1H3/t3-,4-,5+,6-,7-/m1/s1
InChI key
HOVAGTYPODGVJG-XUUWZHRGSA-N
Application
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service