Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

66541075

Sigma-Aldrich

BIONi010-C-11

Human iPS Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509

biological source

human

reprogramming method

episomal

description

age (N/A)

manufacturer/tradename

EBiSC

gender

male

growth mode

adherent (pluripotent)

technique(s)

cell culture | stem cell: suitable

relevant disease(s)

none

shipped in

dry ice

storage temp.

−196°C

General description

Induced pluripotent stem cells (iPSCs) are adult cells that have been reprogrammed to an embryonic stem cell–like state. The cells can replicate indefinitely or, under controlled conditions, can be differentiated into any other cell type such as nerve, heart or liver cells. Medical researchers are able to use iPS cells to test how different patients might respond to new drugs or to analyse how genetic diseases develop.

The EBiSC stem cell bank is a collection of human iPS cells available to academic and commercial researchers for use in disease modelling and other forms of stem cell research. The initial collection has been generated from a wide range of donors representing specific disease backgrounds and healthy controls. EBiSC has established many routine procedures for collecting, expanding and characterizing human iPS cell lines. The stem cell bank includes iPSC cell lines derived from neurodegenerative diseases (Alzheimer′s Disease, Parkinson′s Disease, Dementia, Motor Neuron Disease (ALS) - and Huntington′s Disease), eye and heart diseases, and lines from healthy control donors for age and sex matching.

Cell Line Origin

Depositor
Bioneer

Cell Line Description

Derivation
Primary cell type: -
Reprogramming method
Vector type: Non-integrating
Vector: Episomal
Is the reprogramming vector detectable: Unknown

Xeno free conditions: no
Derived under gmp: no
Available as clinical grade: no

Characterization
Microbiology / Virus Screening
HIV 1: -
HIV 2: -
Hepatitis B: -
Hepatitis C: -
Mycoplasma: Negative

Genotyping
Karyotyping
Passage number: 12
Cell line karyotype: 46, XY
Karyotyping method: G-Banding
Image / Result

Genetic Modification
Disease/phenotype related modifications
Disease: MODY
Type of modification: Isogenic
Gene: HNF1A
Chromosome location: 12q24.31
Nucleotide sequence HGSV: NM_000545.6:c.(c.872dup)
Protein sequence HGSV: NP_000536.5:p.(P291fsinsC)
Zygosity: Homozygous
Target locus modification: Mutated
Description: An additional cytocine has been introduced by CRISPR-Cas9 into a poly-C tract in both alleles of Exon 4 of HNF1A resulting in a out of frame mutation. Western blotting did not show any HNF1A protein in Pancreatic progenitor cells compared to the control line BIONi010-C.

Linkage

Note: EAUA and CLIP must be completed before order fulfillment

Subculture Routine

Surface coating: Matrigel®/Geltrex®
Passage method: Enzyme-free cell dissociation / EDTA
O2 concentration: 20%
CO2 concentration: 5%
Culture medium: mTeSR® 1
Passage Number banked (pre-EBiSC): 12
No. Vials banked (pre-EBiSC): 4

Legal Information

EBiSC is a trademark of Fraunhofer-Gesellschaft
GELTREX is a registered trademark of Life Technologies Corporation
Matrigel is a registered trademark of Corning, Inc.
mTeSR is a registered trademark of WiCell Research Institute, Inc.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service