biological source
human peripheral blood
reprogramming method
Sendai virus
description
age (20-24)
manufacturer/tradename
EBiSC™
gender
female
growth mode
adherent (pluripotent)
technique(s)
cell culture | stem cell: suitable
relevant disease(s)
prolonged QT interval
shipped in
dry ice
storage temp.
−196°C
General description
Induced pluripotent stem cells (iPSCs) are adult cells that have been reprogrammed to an embryonic stem cell–like state. The cells can replicate indefinitely or, under controlled conditions, can be differentiated into any other cell type such as nerve, heart or liver cells. Medical researchers are able to use iPS cells to test how different patients might respond to new drugs or to analyse how genetic diseases develop.
The EBiSC stem cell bank is a collection of human iPS cells available to academic and commercial researchers for use in disease modelling and other forms of stem cell research. The initial collection has been generated from a wide range of donors representing specific disease backgrounds and healthy controls. EBiSC has established many routine procedures for collecting, expanding and characterizing human iPS cell lines. The stem cell bank includes iPSC cell lines derived from neurodegenerative diseases (Alzheimer′s Disease, Parkinson′s Disease, Dementia, Motor Neuron Disease (ALS) - and Huntington′s Disease), eye and heart diseases, and lines from healthy control donors for age and sex matching.
The EBiSC stem cell bank is a collection of human iPS cells available to academic and commercial researchers for use in disease modelling and other forms of stem cell research. The initial collection has been generated from a wide range of donors representing specific disease backgrounds and healthy controls. EBiSC has established many routine procedures for collecting, expanding and characterizing human iPS cell lines. The stem cell bank includes iPSC cell lines derived from neurodegenerative diseases (Alzheimer′s Disease, Parkinson′s Disease, Dementia, Motor Neuron Disease (ALS) - and Huntington′s Disease), eye and heart diseases, and lines from healthy control donors for age and sex matching.
Cell Line Origin
Depositor
Klinikum der Universität zu Köln
Klinikum der Universität zu Köln
Cell Line Description
Derivation
Primary cell type: Peripheral blood mononuclear cell
Tissue collection year: 2017
Passage number reprogrammed: 0
Reprogramming method
Vector type: Non-integrating
Vector: Sendai virus
Gene list:
KLF4
MYC
POU5F1
SOX2
Is the reprogramming vector detectable: No
Merthods used: pcr
Files showing reprogramming vector expressed or silenced: Image / Result
Xeno free conditions: no
Derived under gmp: no
Available as clinical grade: no
Characterization
Analysis of Undifferentiated Cells
Marker expression:
Marker Expressed Immunostaining RT-PCR FACS Enzymatic Assay Expression Profiles
TRA 1-80 Yes - -
FOXD3 Yes -
OCT 4 Yes - -
NANOG Yes - -
DNMT3b Yes -
SOX2 Yes -
SSEA-1 Yes -
SSEA-4 Yes - -
Morphology:
Morphology pictures of the Masterbank and the thawing control do
Morphology pictures of the thawing control d1 and d2
Morphology pictures of the thawing control d3
Differentiation potency
Ectoderm:
Ectoderm
In vitro directed differentiation
Marker Expressed
Nestin Yes
Morphology:
Endoderm:
Endoderm
In vitro directed differentiation
Marker Expressed
SOX17 Yes
Morphology:
Mesoderm:
Mesoderm
In vitro directed differentiation
Marker Expressed
BRACHYURY Yes
Morphology:
Microbiology / Virus Screening
HIV 1: Negative
HIV 2: Negative
Hepatitis B: Negative
Hepatitis C: Negative
Mycoplasma: Negative
Genotyping
Karyotyping
Passage number: 17
Cell line karyotype: No larger chromosomal aberrations to be reported.
Karyotyping method: Molecular karyotyping by SNP array
Image / Result
Primary cell type: Peripheral blood mononuclear cell
Tissue collection year: 2017
Passage number reprogrammed: 0
Reprogramming method
Vector type: Non-integrating
Vector: Sendai virus
Gene list:
KLF4
MYC
POU5F1
SOX2
Is the reprogramming vector detectable: No
Merthods used: pcr
Files showing reprogramming vector expressed or silenced: Image / Result
Xeno free conditions: no
Derived under gmp: no
Available as clinical grade: no
Characterization
Analysis of Undifferentiated Cells
Marker expression:
Marker Expressed Immunostaining RT-PCR FACS Enzymatic Assay Expression Profiles
TRA 1-80 Yes - -
FOXD3 Yes -
OCT 4 Yes - -
NANOG Yes - -
DNMT3b Yes -
SOX2 Yes -
SSEA-1 Yes -
SSEA-4 Yes - -
Morphology:
Morphology pictures of the Masterbank and the thawing control do
Morphology pictures of the thawing control d1 and d2
Morphology pictures of the thawing control d3
Differentiation potency
Ectoderm:
Ectoderm
In vitro directed differentiation
Marker Expressed
Nestin Yes
Morphology:
Endoderm:
Endoderm
In vitro directed differentiation
Marker Expressed
SOX17 Yes
Morphology:
Mesoderm:
Mesoderm
In vitro directed differentiation
Marker Expressed
BRACHYURY Yes
Morphology:
Microbiology / Virus Screening
HIV 1: Negative
HIV 2: Negative
Hepatitis B: Negative
Hepatitis C: Negative
Mycoplasma: Negative
Genotyping
Karyotyping
Passage number: 17
Cell line karyotype: No larger chromosomal aberrations to be reported.
Karyotyping method: Molecular karyotyping by SNP array
Image / Result
Linkage
Note: EAUA and CLIP must be completed before order fulfillment
Subculture Routine
Surface coating: Vitronectin
Passage method: Enzyme-free cell dissociation / EDTA
O2 concentration: 20%
CO2 concentration: 5%
Culture medium: other
Culture base medium: StemFlex™
Main protein source: Albumine
Passage Number banked (pre-EBiSC): 19
No. Vials banked (pre-EBiSC): 25
Passage method: Enzyme-free cell dissociation / EDTA
O2 concentration: 20%
CO2 concentration: 5%
Culture medium: other
Culture base medium: StemFlex™
Main protein source: Albumine
Passage Number banked (pre-EBiSC): 19
No. Vials banked (pre-EBiSC): 25
Legal Information
EBiSC is a trademark of Fraunhofer-Gesellschaft
StemFlex is a trademark of Life Technologies Corporation
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service