Recommended Products
biological source
human
reprogramming method
episomal
description
age (N/A)
manufacturer/tradename
EBiSC™
gender
female
growth mode
adherent (pluripotent)
technique(s)
cell culture | stem cell: suitable
relevant disease(s)
Alzheimer′s disease
shipped in
dry ice
storage temp.
−196°C
General description
The EBiSC stem cell bank is a collection of human iPS cells available to academic and commercial researchers for use in disease modelling and other forms of stem cell research. The initial collection has been generated from a wide range of donors representing specific disease backgrounds and healthy controls. EBiSC has established many routine procedures for collecting, expanding and characterizing human iPS cell lines. The stem cell bank includes iPSC cell lines derived from neurodegenerative diseases (Alzheimer′s Disease, Parkinson′s Disease, Dementia, Motor Neuron Disease (ALS) - and Huntington′s Disease), eye and heart diseases, and lines from healthy control donors for age and sex matching.
Cell Line Origin
Bioneer
Cell Line Description
Primary cell type: -
Reprogramming method
Vector type: Non-integrating
Vector: Episomal
Gene list:
KLF4
Lin28
MYC
POU5F1
SOX2
Is the reprogramming vector detectable: No
Merthods used: pcr
Xeno free conditions: no
Derived under gmp: no
Available as clinical grade: no
Characterization
Microbiology / Virus Screening
HIV 1: Negative
HIV 2: Negative
Hepatitis B: Negative
Hepatitis C: Negative
Mycoplasma: Negative
Genotyping
Karyotyping
Cell line karyotype: 46 XY
Karyotyping method: G-Banding
Image / Result
Genetic Modification
Disease/phenotype related modifications
Disease: Alzheimer′s disease
Type of modification: Isogenic
Gene: Apolipoprotein E
Chromosome location: 19q13.32
Nucleotide sequence HGSV: rs7412 (C/C); rs429358 (C/C)
Zygosity: Homozygous
Target locus modification: The bases at both SNPs rs7412 and rs429358 were changed to a C
Linkage
Subculture Routine
No. Vials banked (pre-EBiSC): 30
Legal Information
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service