biological source
human peripheral blood
reprogramming method
Sendai virus
description
age (50-54)
manufacturer/tradename
EBiSC™
gender
male
growth mode
adherent (pluripotent)
technique(s)
cell culture | stem cell: suitable
relevant disease(s)
familial long QT syndrome
shipped in
dry ice
storage temp.
−196°C
General description
The EBiSC stem cell bank is a collection of human iPS cells available to academic and commercial researchers for use in disease modelling and other forms of stem cell research. The initial collection has been generated from a wide range of donors representing specific disease backgrounds and healthy controls. EBiSC has established many routine procedures for collecting, expanding and characterizing human iPS cell lines. The stem cell bank includes iPSC cell lines derived from neurodegenerative diseases (Alzheimer′s Disease, Parkinson′s Disease, Dementia, Motor Neuron Disease (ALS) - and Huntington′s Disease), eye and heart diseases, and lines from healthy control donors for age and sex matching.
Cell Line Origin
Klinikum der Universität zu Köln
Cell Line Description
Primary cell type: Peripheral blood mononuclear cell
Tissue collection year: 2016
Passage number reprogrammed: 0
Reprogramming method
Vector type: Non-integrating
Vector: Sendai virus
Gene list:
KLF4
MYC
POU5F1
SOX2
Is the reprogramming vector detectable: No
Merthods used: pcr
Notes on reprogramming vector detection: sendai virus absent
Files showing reprogramming vector expressed or silenced: Image / Result
Xeno free conditions: no
Derived under gmp: no
Available as clinical grade: no
Characterization
Analysis of Undifferentiated Cells
Marker expression:
Marker Expressed Immunostaining RT-PCR FACS Enzymatic Assay Expression Profiles
TRA 1-80 Yes - -
SSEA-1 Yes -
NANOG Yes - -
SSEA-4 Yes - -
FOXD3 Yes -
DNMT3b Yes -
OCT 4 Yes - -
Morphology:
pictures of NP0126-1 before bankung and of the thawing control
pictures of NP0126-1 before bankung and of the thawing control
pictures of NP0126-1 before bankung and of the thawing control
Differentiation potency
Ectoderm:
Ectoderm
In vitro directed differentiation
Marker Expressed
Nestin Yes
Morphology:
expression of NESTIN in differentiated iPS cell line NP0126-1
Endoderm:
Endoderm
In vitro directed differentiation
Marker Expressed
SOX17 Yes
Morphology:
expression of SOX17 in differentiated iPS cell line NP0126
Mesoderm:
Mesoderm
In vitro directed differentiation
Marker Expressed
BRACHYURY Yes
Morphology:
expression of BRACHYURY in differentiated iPS cell line NP0126
Microbiology / Virus Screening
HIV 1: Negative
HIV 2: Negative
Hepatitis B: Negative
Hepatitis C: Negative
Mycoplasma: Negative
Sterility
Inoculation for microbiological growth: No Contaminants Detected
Mycoplasma: Not Detected
Viability: Viable post-cryopreservation
Genotyping
Karyotyping
Passage number: 36
Cell line karyotype: No larger chromosomal aberrations observed
Karyotyping method: Molecular karyotyping by SNP array
Image / Result
STR/Fingerprinting: A 16 allele profile has been recorded and data is available upon request, after cell line purchase.
Linkage
Subculture Routine
Passage method: -
Matrix: -
CO2 concentration: -
O2 concentration: -
Temperature: -
Legal Information
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service