Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

66540362

Sigma-Aldrich

BIONi019-A

Human iPS Cell Line

Synonym(s):

EBiSC iPSC Line, Human iPSC, Induced Pluripotent Stem Cell Line, iPS Cell, iPSC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509
Product 66540362 is not currently sold in your country. Contact Technical Service

biological source

human adipose-derived MSC

Quality Level

reprogramming method

episomal

description

age (20-24)

manufacturer/tradename

EBiSC

gender

male

growth mode

adherent (pluripotent)

technique(s)

cell culture | stem cell: suitable

relevant disease(s)

none

shipped in

dry ice

storage temp.

−196°C

General description

Induced pluripotent stem cells (iPSCs) are adult cells that have been reprogrammed to an embryonic stem cell–like state. The cells can replicate indefinitely or, under controlled conditions, can be differentiated into any other cell type such as nerve, heart or liver cells. Medical researchers are able to use iPS cells to test how different patients might respond to new drugs or to analyse how genetic diseases develop.

The EBiSC stem cell bank is a collection of human iPS cells available to academic and commercial researchers for use in disease modelling and other forms of stem cell research. The initial collection has been generated from a wide range of donors representing specific disease backgrounds and healthy controls. EBiSC has established many routine procedures for collecting, expanding and characterizing human iPS cell lines. The stem cell bank includes iPSC cell lines derived from neurodegenerative diseases (Alzheimer′s Disease, Parkinson′s Disease, Dementia, Motor Neuron Disease (ALS) - and Huntington′s Disease), eye and heart diseases, and lines from healthy control donors for age and sex matching.

Cell Line Origin

Depositor
Bioneer

Cell Line Description

Derivation
Primary cell type: Adipose tissue derived mesenchymal stem cell
Passage number reprogrammed: 3
Reprogramming method
Vector type: Non-integrating
Vector: Episomal
Gene list:
KLF4
LIN28
MYC
POU5F1
shP53
SOX2
Is the reprogramming vector detectable: No
Merthods used: pcr
Notes on reprogramming vector detection: DNA isolated at passage 5 or 6 after colony picking. Quantitative PCR with the exogenous pluripotency genes LIN28, SOX2, and OCT4 in iPSC lines. Normal human dermal fibroblasts (NHDFs) transfected two days before with hOSKUL served as a positive control and a sample without template served as negative control.
Vector map: File

Characterization
Analysis of Undifferentiated Cells
Marker expression: SSEA-1(-)SOX2(+)POU5F1 (OCT-4)(+)SSEA-4(+)TDG1(+)NANOG(+)TRA 1-81(+)DMNT3B (+)GABRB3(+)GDF3(+)
Differentiation potency
Ectoderm: In vitro directed differentiation
Marker Expressed:estin (+)Pax6 (+)
Endoderm: In vitro directed differentiation
Marker Expressed:CD184(+)Sox17 (+)
Mesoderm: In vitro directed differentiation
Marker Expressed:CD34(+)CD56(+)

Microbiology / Virus Screening
HIV 1: Negative
HIV 2: Negative
Hepatitis B: Negative
Hepatitis C: Negative
Mycoplasma: Negative
Sterility
Inoculation for microbiological growth: No Contaminants Detected
Mycoplasma: Not Detected
Viability: Viable post-cryopreservation

Genotyping
Karyotyping
Passage number: 11
Cell line karyotype: 46, XY, normal male karyotype
Karyotyping method: G-Banding
Genotyping
STR/Fingerprinting: A 16 allele profile has been recorded and data is available upon request, after cell line purchase.

Linkage

Note: EAUA and CLIP must be completed before order fulfillment

Subculture Routine

Medium: Essential E8®
Passage method: EDTA
Matrix: Matrigel® / Geltrex®
CO2 concentration: 5%
O2 concentration: 5%
Temperature: 37°C

Legal Information

E8 is a registered trademark of WIsconsin Alumni Research Foundation non-stock Corporation
EBiSC is a trademark of Fraunhofer-Gesellschaft
GELTREX is a registered trademark of Life Technologies Corporation
Matrigel is a registered trademark of Corning, Inc.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service