66540013
UKKi007-B
Human iPS Cell Line
Synonym(s):
EBiSC iPSC Line, Human iPSC, Induced Pluripotent Stem Cell Line, iPS Cell, iPSC
About This Item
biological source
human dermis (fibroblast)
Quality Level
reprogramming method
retrovirus
description
age (45-49)
manufacturer/tradename
EBiSC™
gender
female
growth mode
adherent (pluripotent)
technique(s)
cell culture | stem cell: suitable
relevant disease(s)
ventricular tachycardia
shipped in
dry ice
storage temp.
−196°C
General description
The EBiSC stem cell bank is a collection of human iPS cells available to academic and commercial researchers for use in disease modelling and other forms of stem cell research. The initial collection has been generated from a wide range of donors representing specific disease backgrounds and healthy controls. EBiSC has established many routine procedures for collecting, expanding and characterizing human iPS cell lines. The stem cell bank includes iPSC cell lines derived from neurodegenerative diseases (Alzheimer′s Disease, Parkinson′s Disease, Dementia, Motor Neuron Disease (ALS) - and Huntington′s Disease), eye and heart diseases, and lines from healthy control donors for age and sex matching.
Cell Line Origin
Klinikum der Universität zu Köln
Cell Line Description
Primary cell type: Fibroblast of dermis
Primary cell developmental stage: Adult
Location of primary tissue procurement: Berlin, Germany
Tissue collection date: June 15, 2009
Passage number reprogrammed: P3
Reprogramming method
Vector type: Integrating
Vector: Virus
Virus type: Retrovirus
Gene list:
KLF4
MYC
POU5F1
SOX2
Have the reprogramming vectors been silenced: Unknown
Vector map: File
Other:From the same donor: UKKi007-A
Characterization
Analysis of Undifferentiated Cells
Marker expression: POU5F1 (OCT-4)(+)TRA1-80(+)SSEA-4(+)NANOG(+)
Differentiation potency
Ectoderm: In vitro spontaneous differentiation
Marker Expressed:B-TUBULIN(+)FOXG1(-)HES5(+)NEUROD1(+)PAX6 (+)Sox1(+)
Endoderm: In vitro spontaneous differentiation
Marker Expressed:BMP4(+)DCN(+)GATA4(-)HAND1(+)PDGF(+)PECAM (+)VIMENTIN(+)
Mesoderm: In vitro spontaneous differentiation
Marker Expressed:ACTN2, cardiac alpha actinin(+)CXCR4(+)FOXA2(-)GATA6 (+)GSC (+)MLC2v, myosin light chain 2v (+)MYH6, alpha myosin heavy chain(+)PITX1 (+)RYR2 (+)SOX17(+)
Microbiology / Virus Screening
HIV 1: Negative
HIV 2: Not done
Hepatitis B: Negative
Hepatitis C: Negative
Mycoplasma: Negative
Sterility
Inoculation for microbiological growth: No Contaminants Detected
Mycoplasma: Not Detected
Viability: Viable post-cryopreservation
Genotyping
Karyotyping
Karyotyping method: Molecular karyotyping using using OmniExpress Exome Chip
Genotyping
Genome-wide analysis:
SNP-genotyping using OmniExpress Exome Chip
STR/Fingerprinting: A 16 allele profile has been recorded and data is available upon request, after cell line purchase.
Linkage
Subculture Routine
Passage method: EDTA
Matrix: Vitronectin
CO2 concentration: 5%
O2 concentration: 21%
Temperature: 37°C
Legal Information
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service