Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6618

Sigma-Aldrich

Fiske & SubbaRow Reducer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12181503

IVD

for in vitro diagnostic use

Application

For the determination of inorganic phosphorus.

Components

Formulation of: 1-amino-2-naphthol-4-sulfonic acid, sodium sulfite, and bisulfite.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service