Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65986

Sigma-Aldrich

Lipase from Candida antarctica

≥1.0 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

powder

specific activity

≥1.0 U/mg

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Looking for similar products? Visit Product Comparison Guide

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol oleic acid per minute at pH 8.0 and 40 °C (triolein, Cat. No. 62314 as substrate)

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mehran Howaizi et al.
Acta gastro-enterologica Belgica, 75(4), 446-447 (2013-02-14)
Acute pancreatitis has a large number of causes. Major causes are alcohol and gallstones. Toxic causes, mainly represented by medication-induced pancreatitis account for less than 2% of the cases. Cannabis is an anecdotally reported cause of acute pancreatitis. Six cases
Geoff P Lin-Cereghino et al.
Gene, 519(2), 311-317 (2013-03-05)
The methylotrophic yeast, Pichia pastoris, has been genetically engineered to produce many heterologous proteins for industrial and research purposes. In order to secrete proteins for easier purification from the extracellular medium, the coding sequence of recombinant proteins is initially fused
How high-density lipoprotein fuels the failing heart.
Nina Schlueter et al.
Hypertension (Dallas, Tex. : 1979), 61(5), 955-957 (2013-03-06)
First report of telaprevir-induced pancreatitis.
Carlos Ventura et al.
Digestive diseases and sciences, 58(3), 887-888 (2013-02-09)
Leah E Ross et al.
Biochimica et biophysica acta, 1831(6), 1052-1059 (2013-03-09)
The adsorption of colipase is essential for pancreatic triglyceride lipase activity and efficient dietary fat digestion. Yet, little is known about which specific amino acids in the hydrophobic surface of colipase influence adsorption. In this study, we systematically substituted alanine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service