Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

64949

Sigma-Aldrich

7-Methoxycoumarin-3-carboxylic acid N-succinimidyl ester

suitable for fluorescence, ≥97.0% (HPLC)

Synonym(s):

N-Succinimidyl 7-methoxycoumarin-3-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11NO7
CAS Number:
Molecular Weight:
317.25
Beilstein/REAXYS Number:
9145767
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

assay

≥97.0% (HPLC)

solubility

DMF: soluble
acetonitrile: soluble
methanol: soluble

fluorescence

λex 360 nm; λem 410 nm in 0.1 M phosphate pH 7.0

suitability

corresponds for coupling to amines
suitable for fluorescence

storage temp.

−20°C

SMILES string

COc1ccc2C=C(C(=O)ON3C(=O)CCC3=O)C(=O)Oc2c1

InChI

1S/C15H11NO7/c1-21-9-3-2-8-6-10(14(19)22-11(8)7-9)15(20)23-16-12(17)4-5-13(16)18/h2-3,6-7H,4-5H2,1H3

InChI key

JMQAALOXLOSYCQ-UHFFFAOYSA-N

Application

7-Methoxycoumarin-3-carboxylic acid may be used as a reference compound in the purification, separation and analysis of coumarin compounds.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yichao Wang et al.
Analytical biochemistry, 577, 14-20 (2019-04-17)
Insulin is central to regulating fat and carbohydrate metabolism in the body. However, it is difficult to detect insulin using mass spectrometry (MS). The integration of nanotechnology with mass spectrometry for selective and sensitive detection is an important research area.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service