64785
4-Methoxybenzamidine
suitable for fluorescence, ≥96.0% (NT)
Synonym(s):
4-Amidinoanisole
About This Item
assay
≥96.0% (NT)
mp
119-124 °C
fluorescence
λex 393 nm; λem 478 nm (after derivatization with glucose)
λex 393 nm; λem 478 nm(lit.)
suitability
suitable for fluorescence
SMILES string
COc1ccc(cc1)C(N)=N
InChI
1S/C8H10N2O/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3,(H3,9,10)
InChI key
CSISQILZUHMAJB-UHFFFAOYSA-N
Application
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service